Adult Scalp, Unpigmented Slide
# de catalogue 470177-642
Fournisseur: Avantor
Spécifications
- Description:Adult Scalp, Unpigmented
- Cat. no.:470177-642
- Stain:Hematoxylin and Eosin
- Supplier No.:WARD470177-642
- Orientation:Longitudinal section
Spécifications
A propos de cet article
Examine the epidermis of the head
- Identifying features clearly distinguishable
- Rigorous quality control standards
Human. Shows epidermis, hair follicles, sebaceous glands, and other structures.
Customers Who Bought This Also Bought