Human Blood Slide
Catalog # 470177-634
Supplier: Avantor
Specifications
- Description:Human Blood
- Cat. no.:470177-634
- Stain:Giemsa
- Supplier No.:WARD470177-634
- Orientation:Smear
Specifications
About this item
Investigate the components within your blood
- Identifying features clearly distinguishable
- Rigorous quality control standards
Human. Shows the three major functional classes: red cells, white cells and platelets. Human red blood cells have no nuclei.
Customers Who Bought This Also Bought