ContactUsLinkComponent
HUMAN BLOOD (SM) GS SLIDE
Human Blood Slide
Catalog # 470177-634
Supplier:  Avantor
HUMAN BLOOD (SM) GS SLIDE
Human Blood Slide
Catalog # 470177-634
Supplier:  Avantor
Specifications
  • Description:
    Human Blood
  • Cat. no.:
    470177-634
  • Stain:
    Giemsa
  • Supplier No.:
    WARD470177-634
  • Orientation:
    Smear
Specifications
About this item
Investigate the components within your blood
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Human. Shows the three major functional classes: red cells, white cells and platelets. Human red blood cells have no nuclei.
Customers Who Bought This Also Bought