Hyaline Cartilage, Mammal, Trachea
# de catalogue 470177-578
Fournisseur: Avantor
Spécifications
- Description:Hyaline Cartilage, Mammal, Trachea
- Cat. no.:470177-578
- Stain:Hematoxylin and Eosin
- Supplier No.:WARD470177-578
- Orientation:Section
Spécifications
A propos de cet article
Illustrates structural components in the respiratory tract
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal trachea. Small aggregations of chondrocytes.
Customers Who Bought This Also Bought