ContactUsLinkComponent
HYALINE CARTILAGE H/E MAMMAL TRACHEA
Hyaline Cartilage, Mammal, Trachea
# de catalogue 470177-578
Fournisseur:  Avantor
HYALINE CARTILAGE H/E MAMMAL TRACHEA
Hyaline Cartilage, Mammal, Trachea
# de catalogue 470177-578
Fournisseur:  Avantor
Spécifications
  • Description:
    Hyaline Cartilage, Mammal, Trachea
  • Cat. no.:
    470177-578
  • Stain:
    Hematoxylin and Eosin
  • Supplier No.:
    WARD470177-578
  • Orientation:
    Section
Spécifications
A propos de cet article
Illustrates structural components in the respiratory tract
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal trachea. Small aggregations of chondrocytes.
Customers Who Bought This Also Bought