ContactUsLinkComponent
PREPARED SLIDE ROTIFERS WM
Rotifers, Mixed Slide
Catalog # 470177-520
Supplier:  Avantor
PREPARED SLIDE ROTIFERS WM
Rotifers, Mixed Slide
Catalog # 470177-520
Supplier:  Avantor
Specifications
  • Description:
    Rotifers, Mixed Slide
  • Cat. no.:
    470177-520
  • Stain:
    Schneider's and Fast green
  • Supplier No.:
    WARD470177-520
  • Orientation:
    Explano-Mount™
Specifications
About this item
Illustrates the many varieties of Rotifera
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Wheel animacules. Unspecified species.
Customers Who Bought This Also Bought