ContactUsLinkComponent
PREPARED SLIDE MUSHROOM (COPRINUS) CS
Mushroom Anatomy, Coprinus Slide
Catalog # 470177-444
Supplier:  Avantor
PREPARED SLIDE MUSHROOM (COPRINUS) CS
Mushroom Anatomy, Coprinus Slide
Catalog # 470177-444
Supplier:  Avantor
Specifications
  • Description:
    Mushroom Anatomy, Coprinus Slide
  • Cat. no.:
    470177-444
  • Orientation:
    Cross section
  • Stain:
    Quadruple stained
  • Supplier No.:
    WARD470177-444
Specifications
About this item
Investigate mushroom structures
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Entire pileus and gills with basidia and spores.
Customers Who Bought This Also Bought