Mushroom Anatomy, Coprinus Slide
Catalog # 470177-444
Supplier: Avantor
Specifications
- Description:Mushroom Anatomy, Coprinus Slide
- Cat. no.:470177-444
- Orientation:Cross section
- Stain:Quadruple stained
- Supplier No.:WARD470177-444
Specifications
About this item
Investigate mushroom structures
- Identifying features clearly distinguishable
- Rigorous quality control standards
Entire pileus and gills with basidia and spores.
Customers Who Bought This Also Bought