Prepared Nutrient Agar
# de catalogue 470177-374
Fournisseur: IPM SCIENTIFIC, INC.
Numéro CAS:
Spécifications
- Packaging:Plates
- Description:Nutrient Agar
- Cat. no.:470177-374
- Supplier No.:880905
Spécifications
A propos de cet article
General-purpose media support the growth of a variety of bacteria and fungi.
- Non-Selective, General-Purpose Media
- Available as Slant Tubes, Petri Plates, and Heat-and-Pour Bottles
- For Growing a Wide Range of Non-Fastidious Microorganisms
Nutrient agar, a non-selective media, is an excellent choice for growing a wide range of non-fastidious microorganisms. It is frequently used to isolate individual colonies from mixed cultures.
Tube media is configured as 12 tubes per pack. Petri dish media is configured as 10 plates per pack. Pour Paks come with three bottles of media (125 ml) and 20 sterile petri dishes. Just melt and pour.
Customers Who Bought This Also Bought