ContactUsLinkComponent
LM BULLFROG TADPOLES PK/50
Ward's® Live Bullfrog Tadpoles (Rana catesbeiana)
# de catalogue 470177-346
Fournisseur:  Avantor
LM BULLFROG TADPOLES PK/50
Ward's® Live Bullfrog Tadpoles (Rana catesbeiana)
# de catalogue 470177-346
Fournisseur:  Avantor
Spécifications
  • Description:
    Bullfrog Tadpoles, Pack of 50
  • Size:
    At least 2-3"
  • Cat. no.:
    470177-346
  • Supplier No.:
    WARD470177-346
Spécifications
A propos de cet article
Bullfrog tadpoles are great for demonstrating metamorphosis in the classroom.
  • Tadpoles are large
  • Easy to watch as their bodies.
Bullfrog tadpoles take 2 years to develop into frogs. Most tadpoles are supplied 1 year old. Tadpoles are 2 to 3" in size and are available year round.
Ordering information: These products contain living or perishable materials. Please specify a delivery date
Customers Who Bought This Also Bought