Ward's® Live Bullfrog Tadpoles (Rana catesbeiana)
# de catalogue 470177-344
Fournisseur: Avantor
Spécifications
- Description:Bullfrog Tadpoles, Pack of 12
- Size:At least 2-3"
- Cat. no.:470177-344
- Supplier No.:WARD470177-344
Spécifications
A propos de cet article
Bullfrog tadpoles are great for demonstrating metamorphosis in the classroom.
- Tadpoles are large
- Easy to watch as their bodies.
Bullfrog tadpoles take 2 years to develop into frogs. Most tadpoles are supplied 1 year old. Tadpoles are 2 to 3" in size and are available year round.
Ordering information: These products contain living or perishable materials. Please specify a delivery date
Customers Who Bought This Also Bought