Ward's® Live White Planaria (Procotyla fluviatilis)
# de catalogue 470175-822
Fournisseur: Avantor
Spécifications
- Description:Vital Stained White Planaria Culture (Small)
- Storage Conditions:Refrigerator, loose cap
- No. of Students:30
- Cat. no.:470175-822
- Media:Dechlorinated water
- Supplier No.:WARD470175-822
Spécifications
A propos de cet article
Planaria are a flatworm commonly used in science fair projects. They do not bite and move very slowly, making them easy for young students to observe.
- Study structure, feeding habits, responses to environmental stimuli and regeneration
- Compare to Brown and White Planaria (also available)
- Multiple colored specimens in one sample allowing side-by-side comparison
- Vital Stained cultures allow students to better visualize organism structures
- Click on "Product Resources" tab for additional materials for this product
Though smaller than other types of Planaria, this species’ color makes it easier to see its multiple-branched digestive tract.
Vital Stained White Planaria, stained blue, makes these translucent flatworms, and their internal structures, easier to see.
Ordering information: These products contain living or perishable materials.
Delivery: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Customers Who Bought This Also Bought