Ward's® Live Euglena sp. (gracilis)
# de catalogue 470175-818
Fournisseur: Ward's Science
Spécifications
- Description:Euglena gracilis (Small)
- No. of Students:30
- Cat. no.:470175-818
- Packaging:Jar
- Media:Euglena medium
- Supplier No.:WARD470175-818
Spécifications
A propos de cet article
Characterized by a lack of rigid cellulosic walls, instead these organisms have pellicles made of protein. They store paramylon instead of starch.
- Excellent for studying general flagellates
Study Set contains: a culture of Euglena sp. And a Form and Structure Copymaster diagram
Ordering information: These products contain living or perishable materials and will ship via overnight or 2nd day delivery.
Delivery: Please specify a delivery date during checkout.
Customers Who Bought This Also Bought