ContactUsLinkComponent
PIPET BERAL GRAD DSP POLY 150X5MM PK100
Disposable Polyethylene Dropper Pipettes
# de catalogue 470152-908
Fournisseur:  Avantor
PIPET BERAL GRAD DSP POLY 150X5MM PK100
Disposable Polyethylene Dropper Pipettes
# de catalogue 470152-908
Fournisseur:  Avantor
Spécifications
  • Length:
    150 mm
  • Description:
    Beral pipette, graduated, disposable polyethylene, 6572900
  • Cat. no.:
    470152-908
  • Stem Diameter:
    5 mm
  • Supplier No.:
    WARD470152-908
Spécifications
A propos de cet article
Chemically inert polyethylene pipettes.
Designed for consistent bulb pressure and tip circumference to ensure precisely measured drop reproducibility.
Customers Who Bought This Also Bought