Science Kit Polyethylene Specimen Bags
# de catalogue 470151-294
Fournisseur: Avantor
Spécifications
- Description:Bag, specimen, polyethylene, 23×41 cm (12×16"), 4 mil
- Cat. no.:470151-294
- Supplier No.:WARD470151-294
Spécifications
A propos de cet article
Find a multitude of uses for these bags in the laboratory and field.
- Bags can also be heat sealed
Made from clear, 'lay-flat' polyethylene, these bags resist moisture, acids, alkali, embalming fluids, and most solvents and preservatives.
Customers Who Bought This Also Bought