ContactUsLinkComponent
PLATE GLASS HARDNESS 2X3IN PK10
Glass Hardness Plates
# de catalogue 470148-652
Fournisseur:  VWR International
PLATE GLASS HARDNESS 2X3IN PK10
Glass Hardness Plates
# de catalogue 470148-652
Fournisseur:  VWR International
Spécifications
  • Description:
    Glass Hardness Plates, 3x2x1/4"
  • Cat. no.:
    470148-652
  • Supplier No.:
    B19172/1
Spécifications
A propos de cet article
These plates help you determine the hardness of unknown minerals in the field.
  • Plates Determine How Hard Minerals Are
  • System Uses Mohs' Scale
  • Ideal for Unknown Minerals in the Field
  • Custom-Swiped Edges Reduce Risk of Injury
  • Simple-to-Use
Scratch your mineral across the plate determine how hard it is. If the mineral scratches the plates, it’s harder than five on the Mohs’ hardness scale.
The custom-swiped edges reduce risk of accidental injury when handling.
Customers Who Bought This Also Bought