Plastic Scoopula
Catalog # 470125-222
Supplier: Avantor
Specifications
- Description:Scoopula
- Cat. no.:470125-222
- Supplier No.:WARD470125-222
Specifications
About this item
Great for Handling Small Amount of Materials.
- One rounded flared spoon end
- One narrow scoop end
- 8.25" Length
- Polypropylene
Packaging: Scoopulas are available in packs of 20.US Patent 8,709,362
Customers Who Bought This Also Bought