ContactUsLinkComponent
PERANEMA 35 STUDENTS
Ward's® Peranema
# de catalogue 470177-302
Fournisseur:  Ward's Science
PERANEMA 35 STUDENTS
Ward's® Peranema
# de catalogue 470177-302
Fournisseur:  Ward's Science
Spécifications
  • Description:
    Peranema (Small)
  • No. of Students:
    30
  • Cat. no.:
    470177-302
  • Packaging:
    Jar
  • Media:
    Hay culture medium
  • Supplier No.:
    WARD470177-302
Spécifications
A propos de cet article
Extremely small euglenoid. Usually doesn’t rotate when swimming. Scavenger. Hay medium.
Characterized by flagella; lacks plastids.
Customers Who Bought This Also Bought