Ward's® Peranema
# de catalogue 470177-302
Fournisseur: Ward's Science
Spécifications
- Description:Peranema (Small)
- No. of Students:30
- Cat. no.:470177-302
- Packaging:Jar
- Media:Hay culture medium
- Supplier No.:WARD470177-302
Spécifications
A propos de cet article
Extremely small euglenoid. Usually doesn’t rotate when swimming. Scavenger. Hay medium.
Characterized by flagella; lacks plastids.
Customers Who Bought This Also Bought