ContactUsLinkComponent
C. REINHARDTII (+) BROTH 100 STUDENTS
Ward's® Live Chlamydomonas reinhardtii Cultures
Catalog # 470177-288
Supplier:  Avantor
C. REINHARDTII (+) BROTH 100 STUDENTS
Ward's® Live Chlamydomonas reinhardtii Cultures
Catalog # 470177-288
Supplier:  Avantor
Specifications
  • Description:
    C. reinhardtii (+) (Large)
  • Cat. no.:
    470177-288
  • Media:
    Basic culture solution
  • No. of Students:
    100
  • Storage Conditions:
    Room temp, high light, loose cap
  • Packaging:
    Tube
  • Supplier No.:
    WARD470177-288
Specifications
About this item
Demonstrates isogamous reproduction where gametes are similar in appearance and behavior. Make lectures and labs on protist reproduction clearer with our specially selected cultures that illustrate various reproductive phenomona.
  • Both mating types (+) and (-) available
  • Bacteria-free (Axenic) cultures available for both mating types
This green alga is a unicellular flagellate commonly found in damp soil, and aquatic environments. Each cell has 2 flagella and a large eyespot that allow it to sense light.
While all genera of Chlamydomonas are capable of growing in the light, C. reinhartdii can also survive in the dark if it is provided organic carbon.
Genus: Chlamydomonas Species: reinhardtii
Customers Who Bought This Also Bought