ContactUsLinkComponent
DEMO PLATE SORDARIA WILD
Live Sordaria fimicola Cultures
# de catalogue 470177-274
Fournisseur:  Avantor
DEMO PLATE SORDARIA WILD
Live Sordaria fimicola Cultures
# de catalogue 470177-274
Fournisseur:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Sordaria fimicola culture (Wild type)
  • Cat. no.:
    470177-274
  • Packaging:
    Plate
  • Media:
    Sordaria agar
  • Supplier No.:
    WARD470177-274
  • Incubation Conditions:
    30 °C
Spécifications
A propos de cet article
Use Sordaria fimicola to allow students to visualize the results of crossing over during cell division.
  • Use living species to teach complex genetics concepts
  • High quality organisms cultured on-site and shipped to you ready to use
Sordaria's ascospores, exhibit characteristic color patterns based on mutant type and are readily observable under a microscope. Use the arrangement of ascospores to estimate percentage of crossover between the centromere and the gene that controls spore color.
Organisms can be crossed in any confiuration, but the most popular cross (Wild Type strain X Tan Mutant) is available for purchase.
Ordering information: These products contain living or perishable materials. Please specify a delivery date. Allow 2 weeks for delivery. Bacterial and fungal cultures are restricted and can only be ordered by educational and research professionals. Ward's Science is not affiliated with the American Type Culture Collection and does not provide corresponding ATCC numbers for our organisms.
Customers Who Bought This Also Bought