Mammalian Ovary
# de catalogue 470176-970
Fournisseur: Avantor
Spécifications
- Description:Ovary (Mammalian), Developing Follicles
- Cat. no.:470176-970
- Stain:Mallory triple
- Supplier No.:WARD470176-970
- Orientation:Cross Section
Spécifications
A propos de cet article
Observe the graafian follicle in different stages.
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Cat ovary showing graafian follicles. Selected to show germ hill and large ovum.
Ovary (Mammalian), cross section- Demonstrates developing follicles
Ovary (Mammalian), cross section- Shows maturing follicle
Customers Who Bought This Also Bought