Mosquito, Life Cycle Slide
# de catalogue 470176-938
Fournisseur: Avantor
Spécifications
- Description:Mosquito, Life Cycle
- Cat. no.:470176-938
- Stain:unstained
- Supplier No.:WARD470176-938
- Orientation:Whole mount
Spécifications
A propos de cet article
Ectoparasite Life Cycle
- Identifying features clearly distinguishable
- Rigorous quality control standards
The composite includes an egg, larva, pupa, as well as male and female positioned on one slide. Unstained.
Customers Who Bought This Also Bought