ContactUsLinkComponent
MOSQUITO (CULEX SP.) LIFE CYCLE WM
Mosquito, Life Cycle Slide
# de catalogue 470176-938
Fournisseur:  Avantor
MOSQUITO (CULEX SP.) LIFE CYCLE WM
Mosquito, Life Cycle Slide
# de catalogue 470176-938
Fournisseur:  Avantor
Spécifications
  • Description:
    Mosquito, Life Cycle
  • Cat. no.:
    470176-938
  • Stain:
    unstained
  • Supplier No.:
    WARD470176-938
  • Orientation:
    Whole mount
Spécifications
A propos de cet article
Ectoparasite Life Cycle
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
The composite includes an egg, larva, pupa, as well as male and female positioned on one slide. Unstained.
Customers Who Bought This Also Bought