Live Drosophila melanogaster - Standard Crosses, Ward's Science
# de catalogue 470176-774
Fournisseur: Avantor
Spécifications
- Description:LM Drosophila Sepia X Wild (Monohybrid)
- Cat. no.:470176-774
- Supplier No.:WARD470176-774
Spécifications
A propos de cet article
All of our most popular crosses, already done for you. We'll do all the pre-work, so you can spend more time with your students.
- Demonstrate sex-linked, dominant and recessive modes of inheritance
- No pre-work needed - your crosses arrive already done
- Wards offers the shortest lead-time on crosses in the industry, getting your class up and running sooner
- Our cultures come with our satisfaction guarantee for genetic purity and excellent quality
F1 adults will emerge approximately 12 days from date stamped on file.
To create an F2 generation: Once the flies have matured, place five or six sets of males and females into a clean vial with food. You do not need to select virgin females for this step.
Ordering information: For best results, please call our customer service department to place an order for these items.
Customers Who Bought This Also Bought