Ward's® Live Paramecium multimicronucleatum Culture
Catalog # 470176-726
Supplier: Avantor
Specifications
- Description:P. multimicronucleatum (Large)
- Cat. no.:470176-726
- Media:Hay culture medium
- No. of Students:100
- Storage Conditions:Room temp, loose cap
- Packaging:Tube
- Supplier No.:WARD470176-726
Specifications
About this item
Paramecium multimicronucleatumis one of the largest species of Paramecium. This ciliate contains a single macronucleus and four or more micronuclei. Grows in Dilute Alfalfa medium (Hay Medium).
Ordering information: These products contain living or perishable materials and will ship via overnight or 2nd day delivery.
Delivery: Please specify a delivery date during checkout.
Customers Who Bought This Also Bought