ContactUsLinkComponent
PARAMECIUM BURSARIA 35 STUDENTS
Live Paramecium bursaria
# de catalogue 470176-722
PARAMECIUM BURSARIA 35 STUDENTS
Live Paramecium bursaria
# de catalogue 470176-722
Spécifications
  • Description:
    Paramecium bursaria (Small)
  • No. of Students:
    30
  • Cat. no.:
    470176-722
  • Packaging:
    Tube
  • Media:
    Hay culture medium
  • Supplier No.:
    WARD470176-722
Spécifications
A propos de cet article
Paramecium bursaria is a ciliate that contains mutualistic algae called Zoochlorella. The algae lives in the cytoplasma of the Paramecium and provides it with food. Paramecium bursaria grows in Dilute Alfalfa Medium (Hay Medium) or Soil Water Medium. Requires light for growth.
Customers Who Bought This Also Bought