Ward's® Live Blepharisma Culture
# de catalogue 470176-718
Fournisseur: Ward's Science
Spécifications
- Description:Blepharisma (Small)
- No. of Students:30
- Cat. no.:470176-718
- Packaging:Tube
- Media:Hay culture medium
- Supplier No.:WARD470176-718
Spécifications
A propos de cet article
Pink to bright rose color.
Characterized by cilia; nearly all possess two types of nuclei.
- Readyslide available
Vital-Staining is available for this culture; cytoplasmic elements are stained blue on your live culture, to allow for better visibility.
Skip the wet-mount! This organism is available as a set of 5 Readyslides. The tapered end of the container fits directly on your microscope.
Ordering information: These products contain living or perishable materials and will ship via overnight or 2nd day delivery.
Delivery: Please specify a delivery date during checkout.
Customers Who Bought This Also Bought