ContactUsLinkComponent
PEDIASTRUM 35 STUDENTS
Ward's® Pediastrum
# de catalogue 470176-702
Fournisseur:  Ward's Science
PEDIASTRUM 35 STUDENTS
Ward's® Pediastrum
# de catalogue 470176-702
Fournisseur:  Ward's Science
Spécifications
  • Description:
    Pediastrum (Small)
  • No. of Students:
    30
  • Cat. no.:
    470176-702
  • Packaging:
    Tube
  • Media:
    Basic culture solution
  • Supplier No.:
    WARD470176-702
Spécifications
A propos de cet article
Freshwater. Colonial; polygonal cells. Basic culture solution.
Characterized by green plastids and cell walls composed of cellulose.
Customers Who Bought This Also Bought