Ward's® Live Gloeocapsa Culture
# de catalogue 470176-684
Fournisseur: Avantor
Spécifications
- Description:Gloeocapsa (Large)
- Storage Conditions:Room temp, high light, loose cap
- No. of Students:100
- Cat. no.:470176-684
- Packaging:Jar
- Media:Basic culture solution
- Supplier No.:WARD470176-684
Spécifications
A propos de cet article
This colonial cyanobacteria is structured as several cells bound together in a gelatinous "mucilaginous sheath".
Commonly found in masses on wet rocks in aquatic or semi-aquatic habitats.
Genus: Gloeocapsa Species: Not specified
Customers Who Bought This Also Bought