Cosmarium
# de catalogue 470176-678
Fournisseur: Ward's Science
Spécifications
- Description:Cosmarium Culture for 35 Students
- Cat. no.:470176-678
- Supplier No.:WARD470176-678
Spécifications
A propos de cet article
Freshwater. Unicellular; desmid. Basic culture solution.
Characterized by green plastids and cell walls composed of cellulose.
Customers Who Bought This Also Bought