ContactUsLinkComponent
COSMARIUM 35 STUDENTS
Cosmarium
# de catalogue 470176-678
Fournisseur:  Ward's Science
COSMARIUM 35 STUDENTS
Cosmarium
# de catalogue 470176-678
Fournisseur:  Ward's Science
Spécifications
  • Description:
    Cosmarium Culture for 35 Students
  • Cat. no.:
    470176-678
  • Supplier No.:
    WARD470176-678
Spécifications
A propos de cet article
Freshwater. Unicellular; desmid. Basic culture solution.
Characterized by green plastids and cell walls composed of cellulose.
Customers Who Bought This Also Bought