Ward's® Live Chlorella pyrenoidosa Culture
# de catalogue 470176-674
Fournisseur: Ward's Science
Spécifications
- Description:Chlorella pyrenoidosa (Small)
- No. of Students:30
- Cat. no.:470176-674
- Packaging:Tube
- Media:Basic culture solution
- Supplier No.:WARD470176-674
Spécifications
A propos de cet article
This species of freshwater algae is commonly used for research on photosynthesis.
- Excellent for studying photosynthesis
- Rich in cellulose
It is characterized by green plastids and cell walls composed of cellulose. This species has a pyrenoid, which is the organelle that acts as the center of carbon dioxide fixation.
Genus: Chlorella Species: pyrenoidosa
Customers Who Bought This Also Bought