Ward's® Live Oscillatoria Culture
Catalog # 470176-666
Supplier: Avantor
Specifications
- Description:Oscillatoria (Large)
- Storage Conditions:Room temp, high light, loose cap
- No. of Students:100
- Cat. no.:470176-666
- Packaging:Tube
- Media:Basic culture solution
- Supplier No.:WARD470176-666
Specifications
About this item
Oscillatoria is a genus of filamentous green algae that moves by sliding filaments back and forth against each other.
- Filamentous, Freshwater green algae
- Unbranched trichome, highly sheathed
- Genus gets its name from the oscillating motion it exhibits
Genus: Oscillatoria Species: not specified
Frequently Bought Together