Ward's® Live Oscillatoria Culture
Catalog # 470176-664
Supplier: Ward's Science
Specifications
- Description:Oscillatoria (Small)
- Media:Basic culture solution
- Cat. no.:470176-664
- No. of Students:30
- Packaging:Tube
- Supplier No.:WARD470176-664
Specifications
About this item
Oscillatoria is a genus of filamentous green algae that moves by sliding filaments back and forth against each other.
- Filamentous, Freshwater green algae
- Unbranched trichome, highly sheathed
- Genus gets its name from the oscillating motion it exhibits
Genus: Oscillatoria Species: not specified
Customers Who Bought This Also Bought