Scenedesmus
# de catalogue 470176-650
Fournisseur: Ward's Science
Spécifications
- Description:Scenedesmus (Small)
- No. of Students:30
- Cat. no.:470176-650
- Packaging:Tube
- Media:Basic culture solution
- Supplier No.:WARD470176-650
Spécifications
A propos de cet article
Freshwater. Colonial; four cells with spines. Basic culture solution.
Characterized by green plastids and cell walls composed of cellulose.
Customers Who Bought This Also Bought