Hydrodictyon
# de catalogue 470176-640
Fournisseur: Avantor
Spécifications
- Description:Hydrodictyon Culture for 100 Students
- Cat. no.:470176-640
- Supplier No.:WARD470176-640
Spécifications
A propos de cet article
Freshwater. Colonial; net-like structure with multinucleated cells. Basic culture solution.
Characterized by green plastids and cell walls composed of cellulose.
Customers Who Bought This Also Bought