Nostoc, Filamentous
# de catalogue 470176-618
Fournisseur: Ward's Science
Spécifications
- Description:Nostoc, Filamentous (Small)
- No. of Students:30
- Cat. no.:470176-618
- Packaging:Tube
- Media:Basic culture solution
- Supplier No.:WARD470176-618
Spécifications
A propos de cet article
Freshwater. Filamentous. Unbranched, contorted filaments. Basic culture solution.
Shipped in a form that provides the best quality specimen. Most are supplied in 16 x 125 mm tubes, 2 oz. jars, or 8 oz. jars.
Customers Who Bought This Also Bought