Ward's® Live Spirogyra Culture
Catalog # 470176-596
Supplier: Avantor
Specifications
- Description:Spirogyra (Large)
- Cat. no.:470176-596
- Media:Soil water medium
- No. of Students:100
- Storage Conditions:Room temp, high light, loose cap
- Packaging:Jar
- Supplier No.:WARD470176-596
Specifications
About this item
Spirogyra is a genus of filamentous green algae with chloroplasts arranged in a helical (spiral) pattern.
- Excellent food source for freshwater snails and fish
- Undergoes both sexual and asexual reproduction
Spirogyra are a genus of Chlorophyte algae that can be found in freshwater lakes and ponds. They are characterized by green plastids and cell walls composed of cellulose.
Ordering information: Genus: Spirogyra. Species: not specified.
Customers Who Bought This Also Bought