Ward's® Live Freshwater Rotifer Culture
# de catalogue 470176-594
Fournisseur: Avantor
Spécifications
- Description:Freshwater Rotifers (Large culture)
- Storage Conditions:Room temp, loose cap
- No. of Students:100
- Cat. no.:470176-594
- Packaging:3 Tubes
- Media:Hay culture media
- Supplier No.:WARD470176-594
Spécifications
A propos de cet article
Study the Rotifera under your microscope to discover the variety of structures among them. Subculture them in Alfalfa or Hay medium to increase your supply. Species may vary based on availability.
Rotifers have a wide variety of movement types ranging from free moving to sessile. They can be found in any healthy freshwater lakes and ponds.
Customers Who Bought This Also Bought