Ward's® Live Paramecium caudatum Culture
# de catalogue 470176-582
Fournisseur: Avantor
Spécifications
- Description:Paramecium caudatum, for 120 students
- No. of Students:100
- Cat. no.:470176-582
- Packaging:Tube (3)
- Media:Hay culture medium
- Supplier No.:WARD470176-582
Spécifications
A propos de cet article
Paramecium caudatum is a common protist that inhabits a variety of aquatic ecosystems.
- Genus: Paramecium
- Species: caudatum
- Common Protist That Inhabits a Variety of Aquatic Ecosystems
Paramecium are characterized by having two nuclei (a large macro and smaller micronucleus). They feed on bacteria and other microscopic organisms. Paramecium are a common food source for other protozoa such as Didinium and Chaos.
They are covered with tiny "hair-like" organelles called cilia. These cilia are used in both feeding and locomotion.
Customers Who Bought This Also Bought