Ward's® Live Chaos (Pelomyxa sp.) Culture
# de catalogue 470176-574
Fournisseur: Avantor
Spécifications
- Description:Pelomyxa Carolinensis - Chaos (Small)
- No. of Students:30
- Cat. no.:470176-574
- Packaging:Jar
- Media:Distilled water
- Supplier No.:WARD470176-574
Spécifications
A propos de cet article
Members of the Pelomyxa genus look very much like Amoeba. Pelomyxa, however, have many nuclei, whereas Amoeba have only one.
Vital-Staining is available for this culture; cytoplasmic elements are stained blue on your live culture, to allow for better visibility.
Ordering information: These products contain living or perishable materials and will ship via overnight or 2nd day delivery.
Delivery: Please specify a delivery date during checkout.
Customers Who Bought This Also Bought