ContactUsLinkComponent
LM BURSARIA TRUNCATELLA 30 STUD
Ward's® Live Bursaria truncatella Culture
# de catalogue 470176-566
Fournisseur:  Ward's Science
LM BURSARIA TRUNCATELLA 30 STUD
Ward's® Live Bursaria truncatella Culture
# de catalogue 470176-566
Fournisseur:  Ward's Science
Spécifications
  • Description:
    Bursaria truncatella (Small)
  • No. of Students:
    30
  • Cat. no.:
    470176-566
  • Packaging:
    Jar
  • Media:
    Distilled water
  • Supplier No.:
    WARD470176-566
Spécifications
A propos de cet article
Very large ciliate. Feeds on colpidium. Hay medium.
Characterized by cilia; nearly all possess two types of nuclei.
Ordering information: These products contain living or perishable materials
Delivery: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Customers Who Bought This Also Bought