Ward's® Live Bursaria truncatella Culture
# de catalogue 470176-566
Fournisseur: Ward's Science
Spécifications
- Description:Bursaria truncatella (Small)
- No. of Students:30
- Cat. no.:470176-566
- Packaging:Jar
- Media:Distilled water
- Supplier No.:WARD470176-566
Spécifications
A propos de cet article
Very large ciliate. Feeds on colpidium. Hay medium.
Characterized by cilia; nearly all possess two types of nuclei.
Ordering information: These products contain living or perishable materials
Delivery: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Customers Who Bought This Also Bought