Ward's® Live Brown Planaria (Dugesia dorotocephala)
# de catalogue 470176-558
Fournisseur: Ward's Science
Spécifications
- Description:Brown Planaria
- Storage Conditions:Refrigerator, loose cap
- No. of Students:35
- Cat. no.:470176-558
- Media:Dechlorinated water
- Supplier No.:WARD470176-558
Spécifications
A propos de cet article
This is the most widely studied Turbellarian, a free-living flatworm commonly used in science fair projects.
- Students Research Structure, Feeding Habits, Responses to Environmental Stimuli, and Regeneration
- Easy-To-Maintain
- Planaria Feed on Animal Matter
- Water Must Be Changed After Feeding
Brown planaria are easy to maintain. They feed on animal matter such as liver and are up to 1/4" in size.
Ordering information: These products contain living or perishable materials. Please specify a delivery date.
Customers Who Bought This Also Bought