Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Spécifications
- Description:Bacillus subtilis
- Cat. no.:470176-524
- Packaging:Tube
- Media:Tryptic soy agar
- Supplier No.:WARD470176-524
- Incubation Conditions:30 °C
Spécifications
À propos de cet article
This organism is commonly isolated in soil and produces antibiotics such as bacitracin, subtilisin, and mycobacillin.
- Biosafety Level: 1
- Gram Stain: Positive
- Morphology: rods in chains
- Motility: Positive
- Respiration: Aerobic
- Spores: Yes, centrally located
All bacterial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
It's gram stain is positive, and its spores are centrally located.