Ward's® Live Anoles (Anolis carolinensis)
# de catalogue 470176-520
Fournisseur: CYRS BIOLOGY CO.
Spécifications
- Description:Anoles
- Cat. no.:470176-520
- Body Size:5 - 6"
- Supplier No.:WARD470176-520
Spécifications
A propos de cet article
Anoles (American Chameleons) are tree-dwelling lizards found mostly in the southeastern US.
- Their ability to change color makes them excellent for discussions about camouflage and adaptation
- Watch their mating and feeding behaviors
- Easy to care for
In addition to the fact that they change colors, Anoles are interesting creatures to watch because they are extremely territorial. Males have a dewlap that they use for display purposes; when defending his territory.
They require a heat source to maintain a steady temperature at 72 to 80°F and direct light.
Ordering information: These products contain living or perishable materials
Delivery: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Customers Who Bought This Also Bought