ContactUsLinkComponent
PHILODINA 35 STUDENTS
Ward's® Philodina
# de catalogue 470176-460
Fournisseur:  Ward's Science
PHILODINA 35 STUDENTS
Ward's® Philodina
# de catalogue 470176-460
Fournisseur:  Ward's Science
Spécifications
  • Description:
    Philodina, For 35 Students
  • Cat. no.:
    470176-460
  • Supplier No.:
    WARD470176-460
Spécifications
A propos de cet article
Philodina sp. Easy to culture and easy to examine under a microscope because of the lack of a lorica (shell), these typical rotifers are perfect for general study. Culture instructions are included.
Customers Who Bought This Also Bought