Ward's® Philodina
# de catalogue 470176-460
Fournisseur: Ward's Science
Spécifications
- Description:Philodina, For 35 Students
- Cat. no.:470176-460
- Supplier No.:WARD470176-460
Spécifications
A propos de cet article
Philodina sp. Easy to culture and easy to examine under a microscope because of the lack of a lorica (shell), these typical rotifers are perfect for general study. Culture instructions are included.
Customers Who Bought This Also Bought