ContactUsLinkComponent
P. CAUDATUM VITAL STAIN 35 STUDENTS
Ward's® Live Paramecium caudatum Culture
# de catalogue 470176-450
Fournisseur:  Avantor
Ward's® Live Paramecium caudatum Culture
# de catalogue 470176-450
Fournisseur:  Avantor
Spécifications
  • Description:
    Paramecium caudatum, VITAL-STAIN 35 Students
  • No. of Students:
    35
  • Cat. no.:
    470176-450
  • Packaging:
    Tube (1)
  • Media:
    Hay culture medium
  • Supplier No.:
    WARD470176-450
Spécifications
A propos de cet article
Paramecium caudatum is a common protist that inhabits a variety of aquatic ecosystems.
  • Genus: Paramecium
  • Species: caudatum
  • Common Protist That Inhabits a Variety of Aquatic Ecosystems
Paramecium are characterized by having two nuclei (a large macro and smaller micronucleus). They feed on bacteria and other microscopic organisms. Paramecium are a common food source for other protozoa such as Didinium and Chaos.
They are covered with tiny "hair-like" organelles called cilia. These cilia are used in both feeding and locomotion. 
Customers Who Bought This Also Bought