ContactUsLinkComponent
CHAOS VITAL STAIN 35 STUDENTS
Ward's® Live Chaos (Pelomyxa sp.) Culture
# de catalogue 470176-448
Fournisseur:  Avantor
CHAOS VITAL STAIN 35 STUDENTS
Ward's® Live Chaos (Pelomyxa sp.) Culture
# de catalogue 470176-448
Fournisseur:  Avantor
Spécifications
  • Description:
    Vital Stained Chaos (Small)
  • Storage Conditions:
    Room temp, loose cap
  • No. of Students:
    30
  • Cat. no.:
    470176-448
  • Packaging:
    Jar
  • Media:
    Distilled water
  • Supplier No.:
    WARD470176-448
Spécifications
A propos de cet article
Members of the Pelomyxa genus look very much like Amoeba. Pelomyxa, however, have many nuclei, whereas Amoeba have only one.
Vital-Staining is available for this culture; cytoplasmic elements are stained blue on your live culture, to allow for better visibility.
Ordering information: These products contain living or perishable materials and will ship via overnight or 2nd day delivery.
Delivery: Please specify a delivery date during checkout.
Customers Who Bought This Also Bought