Ward's® Live Arcella Culture
Catalog # 470176-436
Supplier: Ward's Science
Specifications
- Description:Arcella vulgaris (Small)
- Cat. no.:470176-436
- Media:Amoeba medium
- No. of Students:30
- Packaging:Jar
- Supplier No.:WARD470176-436
Specifications
About this item
Usually found in freshwater and mosses, this organism has a hard umbrella shaped shell or "test", out of which finger-like pseudopodia project.
- Contains a "Test"
- Pseudopodia
Ordering information: These products contain living or perishable materials and will ship via overnight or 2nd day delivery.
Delivery: Please specify a delivery date during checkout.
Customers Who Bought This Also Bought