Ward's® Peridinium
Catalog # 470176-432
Supplier: Ward's Science
Specifications
- Description:Peridinium (Small)
- Cat. no.:470176-432
- Media:Soil water medium
- No. of Students:30
- Packaging:Jar
- Supplier No.:WARD470176-432
Specifications
About this item
Freshwater. Unicellular. Soil-water medium. Characterized by two flagella (one inside a groove at right angles to the other); most also have tests made of cellulose and are encrusted with silica.
Customers Who Bought This Also Bought