ContactUsLinkComponent
AMPHIDINIUM 35 STUDENTS
Ward's® Live Amphidinium Culture
# de catalogue 470176-428
Fournisseur:  Ward's Science
Ward's® Live Amphidinium Culture
# de catalogue 470176-428
Fournisseur:  Ward's Science
Spécifications
  • Description:
    Amphidinium (Small)
  • No. of Students:
    30
  • Cat. no.:
    470176-428
  • Packaging:
    Tube
  • Media:
    Erdschreiber's medium
  • Supplier No.:
    WARD470176-428
Spécifications
A propos de cet article
Marine; Small Motile Cells with the Transverse Furrow Located Near the Apex.
Ordering information: These products contain living or perishable materials and will ship via overnight or 2nd day delivery.
Delivery: Please specify a delivery date during checkout.
Customers Who Bought This Also Bought