Ward's® Live Amphidinium Culture
# de catalogue 470176-428
Fournisseur: Ward's Science
Spécifications
- Description:Amphidinium (Small)
- No. of Students:30
- Cat. no.:470176-428
- Packaging:Tube
- Media:Erdschreiber's medium
- Supplier No.:WARD470176-428
Spécifications
A propos de cet article
Marine; Small Motile Cells with the Transverse Furrow Located Near the Apex.
Ordering information: These products contain living or perishable materials and will ship via overnight or 2nd day delivery.
Delivery: Please specify a delivery date during checkout.
Customers Who Bought This Also Bought