Synura
# de catalogue 470176-424
Fournisseur: Ward's Science
Spécifications
- Description:Synura, for 30 Students
- Cat. no.:470176-424
- Supplier No.:WARD470176-424
Spécifications
A propos de cet article
Freshwater; elongated, biflagellate cells that form globular colonies; gelatinous sheath absent.
Live Cultures are supplied with media in 16 x 125 mm tubes, 2 oz. jars, or 8 oz. jars. Fast delivery is available directly from our laboratory. Please specify a delivery date.
Each culture shipment is prepared in our laboratories and includes the "Working with Algae & Cyanobacteria" manual which contains detailed instructions for subculturing and use.
Canadian Customers:
CFIA and PHAC regulations require customers to submit a completed inspection checklist and signed statement of facilities compliance before receiving certain cultures. A customer service representative will contact you if documentation is required.
Customers Who Bought This Also Bought