Synedra
# de catalogue 470176-422
Fournisseur: Ward's Science
Spécifications
- Description:Synedra (Small)
- No. of Students:30
- Cat. no.:470176-422
- Packaging:Tube
- Media:Basic culture solution
- Supplier No.:WARD470176-422
Spécifications
A propos de cet article
Freshwater, Unicellular; pennate, with bilateral symmetry. Diatom or basic medium.
Characterized by shells (tests) made of silica, in two parts.
Customers Who Bought This Also Bought