ContactUsLinkComponent
SYNEDRA 35 STUDENTS
Synedra
# de catalogue 470176-422
Fournisseur:  Ward's Science
SYNEDRA 35 STUDENTS
Synedra
# de catalogue 470176-422
Fournisseur:  Ward's Science
Spécifications
  • Description:
    Synedra (Small)
  • No. of Students:
    30
  • Cat. no.:
    470176-422
  • Packaging:
    Tube
  • Media:
    Basic culture solution
  • Supplier No.:
    WARD470176-422
Spécifications
A propos de cet article
Freshwater, Unicellular; pennate, with bilateral symmetry. Diatom or basic medium.
Characterized by shells (tests) made of silica, in two parts.
Customers Who Bought This Also Bought