Dictyostelium discoideum, Tube Culture
Catalog # 470176-392
Supplier: Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Dictyostelium discoideum
- Cat. no.:470176-392
- Media:Cornmeal agar
- No. of Students:30
- Packaging:Tube
- Supplier No.:WARD470176-392
Specifications
About this item
Cellular slime mold. Demonstrates amoeboid state, pseudoplasmodial state, and fruiting body, or sorocarp.
- Found in woodland soil in decomposing vegetation where bacteria are abundant
- Characterized by an amoeboid stage (lacking cell walls) and a fungus-like reproductive structure
- Incubation temperature 25°C
Customers Who Bought This Also Bought