Spécifications
- Description:Tribonema, for 30 Students
- Cat. no.:470176-364
- Supplier No.:WARD470176-364
Spécifications
A propos de cet article
Freshwater; unbranched filaments with cylindrical- to slightly barrel-shaped cells; shows the characteristic H-pieces upon fragmentation of the filament.
Live Cultures are supplied with media in 16 x 125 mm tubes, 2 oz. jars, or 8 oz. jars. Fast delivery is available directly from our laboratory. Please specify a delivery date.
Each culture shipment is prepared in our laboratories and includes the "Working with Algae & Cyanobacteria" manual which contains detailed instructions for subculturing and use.
Canadian Customers:
CFIA and PHAC regulations require customers to submit a completed inspection checklist and signed statement of facilities compliance before receiving certain cultures. A customer service representative will contact you if documentation is required.